H-Lys(Tfa)-NCA ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:42267-27-6
:MFCD20231371
Formula:C₉H₁₁F₃N₂O₄
MDL Number:MFCD11046652
:268.2 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Specific rotation [a]20/D:-30.0 to -34.0 deg (C=3, Dioxane)
Specification Test Results
Frequently Bought Together