
2-(2,6-Diisopropylphenyl)-3,3-dimethyl-2-azaspiro[4.5]dec-1-en-2-ium Hydrogen Dichloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1141464-90-5
:
Formula:C₂₃H₃₇Cl₂N
MDL Number:MFCD20922893
:398.46 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together