Order Entry
Netherlands
ContactUsLinkComponent
Dinaphtho[2,1-b:1',2'-d]furan ≥98.0% (by GC)
Dinaphtho[2,1-b:1',2'-d]furan ≥98.0% (by GC)
Supplier:  TCI
Certificates
Dinaphtho[2,1-b:1',2'-d]furan ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    157.0 to 161.0 deg-C

Specification Test Results

Frequently Bought Together