
3-(4,4-difluoro-5,7-dimethyl-4-bora-3a,4a-diaza-s-indacene-3-yl)propionic acid NHS ester ≥98.0% (by HPLC)
: TCI
:146616-66-2
:
:C₁₈H₁₈BF₂N₃O₄
:MFCD00467385
:389.166 g/mol
:GHS07
:Freezer
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:160.0 to 164.0 deg-C
- Elemental analysis (Nitrogen):10.30 to 11.40 %
Frequently Bought Together