5-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-yl)-2,3-dihydrothieno[3,4-b][1,4]dioxane ≥98.0% (by GC)
Catalog # TCIAD5525-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:91.0 to 95.0 deg-C
Specification Test Results
About this item
:
:268.14 g/mol
Storage Temperature:Refrigerator
CAS Number:250726-93-3
Formula:C₁₂H₁₇BO₄S
MDL Number:MFCD08060541