
1,1,3,3-Tetramethoxypropane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:102-52-3
:
Formula:C₇H₁₆O₄
MDL Number:MFCD00008488
:164.2 g/mol
Boiling Pt:183 °C (760 mmHg)
Density:0.99 g/cm³
Flash Pt:54 °C
Storage Temperature:Ambient
EINECS:203-037-2
UN:3271
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4050 to 1.4090 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Related Information
Frequently Bought Together