Order Entry
Netherlands
ContactUsLinkComponent
 
 
1,3-Diisopropylimidazolium hydrogencarbonate (contains varying amounts of 1,3-Diisopropylimidazolium-2-carboxylate)
  TCIAD5498-1G
 :  TCI
 :  
 
1,3-Diisopropylimidazolium hydrogencarbonate (contains varying amounts of 1,3-Diisopropylimidazolium-2-carboxylate)
  TCIAD5498-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    97.0 to 103.0 %

 

 

 :
 :214.27 g/mol
 :GHS07
 :1372124-90-7
 :C₁₀H₁₈N₂O₃