Order Entry
Netherlands
ContactUsLinkComponent
3,3'''-Dihexyl-2,2':5',2'':5'',2'''-quaterthiophene ≥95.0% (by GC) (contains 3% Dichloromethane at maximum)
Health
3,3'''-Dihexyl-2,2':5',2'':5'',2'''-quaterthiophene ≥95.0% (by GC) (contains 3% Dichloromethane at maximum)
Catalog # TCIAD5492-200MG
Supplier:  TCI
CAS Number:  
Health
3,3'''-Dihexyl-2,2':5',2'':5'',2'''-quaterthiophene ≥95.0% (by GC) (contains 3% Dichloromethane at maximum)
Catalog # TCIAD5492-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid to cloudy liquid
  • Purity (GC):
    min. 95.0 %
  • Methylene chloride (NMR):
    max. 3.0 %

Specification Test Results

About this item

 :
 :498.82 g/mol
 :GHS08
Storage Temperature:Refrigerator
CAS Number:132814-91-6
Formula:C₂₈H₃₄S₄
MDL Number:MFCD10567045