
3,4-Diethoxyaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:39052-12-5
:
Formula:C₁₀H₁₅NO₂
MDL Number:MFCD00026782
:181.24 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:46.0 to 50.0 deg-C
Specification Test Results
Frequently Bought Together