
1,3-Dimethylimidazolium-2-carboxylate ≥98.0% (by titrimetric analysis)
Catalog # TCIAD5396-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g/mol
- Seq:9220
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
:140.14 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:536755-29-0
Formula:C₆H₈N₂O₂
MDL Number:MFCD07636581