Order Entry
Netherlands
ContactUsLinkComponent
6,6'-Dibromodi(2-octyldodecyl)isoindigo ≥99.0% (by HPLC)
6,6'-Dibromodi(2-octyldodecyl)isoindigo ≥99.0% (by HPLC)
Catalog # TCIAD5386-200MG
Supplier:  TCI
CAS Number:  
6,6'-Dibromodi(2-octyldodecyl)isoindigo ≥99.0% (by HPLC)
Catalog # TCIAD5386-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Yellow to Amber to Dark red powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    51.0 to 55.0 deg-C

Specification Test Results

About this item

 :
 :981.14 g/mol
CAS Number:1263379-85-6
Formula:C₅₆H₈₈Br₂N₂O₂