4,4'-Dibromo-4''-(hexyloxy)triphenylamine ≥97.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:1456880-51-5
:
Formula:C₂₄H₂₅Br₂NO
:503.28 g/mol
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid to slightly cloudy liquid
- Purity (HPLC):min. 95.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
Specification Test Results
Frequently Bought Together