6,6'-Diazido-6,6'-dideoxytrehalose ≥93.0% (by HPLC)
Catalog # TCIAD5372-50MG
Supplier: TCI
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC) (CAD):min. 93.0 area%
- Elemental analysis (Nitrogen):20.00 to 23.00 %
Specification Test Results
About this item
:
:392.33 g/mol
Melting Pt:211°C
Storage Temperature:Refrigerator
CAS Number:18933-88-5
Formula:C₁₂H₂₀N₆O₉