9,9-Di-n-octylfluorene-2-boronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD5371-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:57.0 to 61.0 deg-C
Specification Test Results
About this item
:
:516.61 g/mol
CAS Number:302554-81-0
:MFCD20926226
Formula:C₃₅H₅₃BO₂
MDL Number:MFCD16294553