
1-Dodecyl-3-methylimidazolium bromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:61546-00-7
:
Formula:C₁₆H₃₁BrN₂
MDL Number:MFCD17014729
:331.3347 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together