
1,3-Dimethylimidazolium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:174899-81-1
:
Formula:C₇H₉F₆N₃O₄S₂
MDL Number:MFCD18251322
:377.28 g/mol
Melting Pt:22°C
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to lump
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):10.50 to 11.50 %
Specification Test Results
Frequently Bought Together