Order Entry
Netherlands
ContactUsLinkComponent
1,3-Dihydroxy-9H-xanthen-9-one ≥98.0% (by HPLC)
1,3-Dihydroxy-9H-xanthen-9-one ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
1,3-Dihydroxy-9H-xanthen-9-one ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Elemental analysis (Carbon):
    66.00 to 71.00 %

Specification Test Results

Frequently Bought Together