2-Dodecylhexadecan-1-ol ≥93.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:72388-18-2
:
Formula:C₂₈H₅₈O
MDL Number:MFCD27665210
:410.77 g/mol
Storage Temperature:Refrigerator
EINECS:276-627-0
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 93.0 %
Specification Test Results
Frequently Bought Together