
Diethyl [2,2'-Bipyridine]-5,5'-dicarboxylate ≥98.0% (by GC, titration analysis)
: TCI
:1762-46-5
:
:C₁₆H₁₆N₂O₄
:MFCD11110569
:300.31 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:148.0 to 152.0 deg-C
Frequently Bought Together