Order Entry
Netherlands
ContactUsLinkComponent
 
1,6-Di-O-acetyl-2,3,4-tri-O-benzyl-alpha-D-mannopyranose ≥95.0% (by HPLC)
Catalog # TCIAD5294-1G
Supplier:  TCI
CAS Number:  
1,6-Di-O-acetyl-2,3,4-tri-O-benzyl-alpha-D-mannopyranose ≥95.0% (by HPLC)
Catalog # TCIAD5294-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 95.0 area%

Specification Test Results

About this item

 :
 :534.61 g/mol
Melting Pt:96°C
Storage Temperature:Freezer
CAS Number:65556-30-1
Formula:C₃₁H₃₄O₈