Order Entry
Netherlands
ContactUsLinkComponent
1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane ≥96.0% (by GC)
Irritant
1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane ≥96.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Melting point:
    48.0 to 52.0 deg-C

Specification Test Results

Frequently Bought Together