5,10-Dibromonaphtho[1,2-c:5,6-c']bis([1,2,5]thiadiazole) ≥96.0% (by HPLC)
Catalog # TCIAD5288-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 96.0 area%
- Elemental analysis (Nitrogen):13.20 to 14.60 %
Specification Test Results
About this item
:
:402.08 g/mol
CAS Number:133546-50-6
Formula:C₁₀H₂Br₂N₄S₂