
2,4-Dibromomesitylene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6942-99-0
:
Formula:C₉H₁₀Br₂
MDL Number:MFCD00013525
:277.99 g/mol
Boiling Pt:279°C
EINECS:230-100-1
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:61.0 to 65.0 deg-C
Specification Test Results
Frequently Bought Together