
1,3-Diaminopropane dihydrochloride, low water content ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:10517-44-9
:
Formula:C₃H₁₂Cl₂N₂
MDL Number:MFCD00012525
:147.05 g/mol
EINECS:234-051-7
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:250.0 to 255.0 deg-C
- Water:max. 100 ppm
Specification Test Results
Frequently Bought Together