
2-(5,6-Dimethyl-9-oxoxanthen-4-yl)acetic acid (DMXAA) ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:117570-53-3
:
Formula:C₁₇H₁₄O₄
MDL Number:MFCD00870555
:282.295 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Frequently Bought Together