
4-(Dimethylcarbamoyl)phenylboronic acid ≥97.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:405520-68-5
:
Formula:C₉H₁₂BNO₃
MDL Number:MFCD02258943
:193.01 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 110.0 %
Specification Test Results
Frequently Bought Together