
4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane ≥95.0% (by GC)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:57280-22-5
:
Formule:C₇H₁₂O₃
MDL-nummer:MFCD16621162
:144.17 g/mol
Kokend Pt:86°C
EINECS:421-750-9
:GHS07
Temperatuur bij opslag:Refrigerator
Specificatie testresultaten
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specificatie testresultaten
Frequently Bought Together