
1,3-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester ≥97.0% (by GC, titration analysis)
Catalog # TCIAD5184-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:73.0 to 77.0 deg-C
Specification Test Results
About this item
:
:222.0918 g/mol
:GHS07
CAS Number:1046832-21-6
Formula:C₁₁H₁₉BN₂O₂
MDL Number:MFCD09864190