
4,7-Dihydroxycoumarin ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1983-81-9
:
Formula:C₉H₆O₄
MDL Number:MFCD00203882
:178.14 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together