
2,6-Dibromo-3-hydroxypyridine ≥97.0% (by titrimetric analysis)
Catalog # TCIAD5174-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:164.0 to 168.0 deg-C
Specification Test Results
About this item
:
:252.8900 g/mol
EINECS:229-548-0
:GHS07
CAS Number:6602-33-1
Formula:C₅H₃Br₂NO
MDL Number:MFCD01318479