Order Entry
Netherlands
ContactUsLinkComponent
Disodium 3,3'-Dithiobis(1-propanesulphonate) ≥97.0% (by titrimetric analysis)
Irritant
Disodium 3,3'-Dithiobis(1-propanesulphonate) ≥97.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Disodium 3,3'-Dithiobis(1-propanesulphonate) ≥97.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Ion exchange titration):
    min. 97.0 %

Specification Test Results

Frequently Bought Together