
(3,5-Dichlorophenyl)(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulphonate ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1868173-25-4
:
Formula:C₂₂H₂₁Cl₂IO₆S
MDL Number:MFCD30475653
:611.27 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 95.0 % (total of Ion-Pair)
Specification Test Results
Frequently Bought Together