Order Entry
Netherlands
ContactUsLinkComponent
Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine ≥97.0% (by GC)
Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine ≥97.0% (by GC)
Supplier:  TCI
Certificates
undefined
Dicyclohexyl(2,4,6-triisopropylphenyl)phosphine ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    137.0 to 141.0 deg-C

Specification Test Results

Frequently Bought Together