1,2-Difluoro-4-[trans-4-[2-(trans-4-propylcyclohexyl)ethyl]cyclohexyl]benzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:117943-37-0
:
Formula:C₂₃H₃₄F₂
MDL Number:MFCD12912373
:348.52 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Mesomorphic range:37 to 107 deg-C
Specification Test Results
Frequently Bought Together