
6-Phenyl-4,5-dihydropyridazin-3(2H)-one ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:1011-46-7
:
Formula:C₁₀H₁₀N₂O
MDL Number:MFCD00099597
:174.2 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:150.0 to 154.0 deg-C
Specification Test Results
Frequently Bought Together