
Docosyl acrylate ≥95.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:18299-85-9
:
Formula:C₂₅H₄₈O₂
MDL Number:MFCD00080409
:380.66 g/mol
:Stabilised with 4-methoxyphenol (MEHQ)
EINECS:242-182-6
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 95.0 %
- Melting point:45.0 to 49.0 deg-C
Specification Test Results
Frequently Bought Together