
3,5-Di-tert-butylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:197223-39-5
:
Formula:C₁₄H₂₃BO₂
MDL Number:MFCD07780212
:234.15 g/mol
Melting Pt:182°C
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):97.0 to 108.0 %
Specification Test Results
Frequently Bought Together