
2,7-Dibromo-9,9-difluorofluorene ≥97.0% (by GC)
Catalog # TCIAD5053-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:138.0 to 142.0 deg-C
Specification Test Results
About this item
:
:360.00 g/mol
:GHS07
CAS Number:1229603-71-7
Formula:C₁₃H₆Br₂F₂
MDL Number:MFCD28386928