
2-Dicyclohexylphosphino-2',6'-diisopropoxybiphenyl ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:787618-22-8
:
Formula:C₃₀H₄₃O₂P
MDL Number:MFCD06798294
:466.65 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:123.0 to 127.0 deg-C
Specification Test Results
Frequently Bought Together