
Dimethyl 2,2'-Bipyridine-6,6'-dicarboxylate ≥95.0% (by GC)
: TCI
:142593-07-5
:
:C₁₄H₁₂N₂O₄
:MFCD11042478
:272.26 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:gt. 192.0 and le 196.0 deg-C
Frequently Bought Together