Order Entry
Netherlands
ContactUsLinkComponent
3',5'-Dichloro-4'-Hydroxyacetophenone ≥98.0% (by GC, titration analysis)
Irritant
3',5'-Dichloro-4'-Hydroxyacetophenone ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
3',5'-Dichloro-4'-Hydroxyacetophenone ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    162.0 to 166.0 deg-C

Specification Test Results

Frequently Bought Together