
1,1'-Dibutyl-3,3,3',3'-tetramethylindotricarbocyanine Hexafluorophosphate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:134339-08-5
:
Formula:C₃₅H₄₅F₆N₂P
:638.72 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Lambda max.:743.0 to 748.0 nm (MeOH)
- Absorbance (E1%1cm):min. 4200 (MeOH, 743 to 748nm)
Specification Test Results
Frequently Bought Together