4,7-Dibromo-1,10-phenanthroline hydrate ≥98.0% (by HPLC)
: TCI
:156492-30-7
:
:C₁₂H₆Br₂N₂·xH₂O
:MFCD25562940
:338.00 g/mol
:236°C
- Appearance:White to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 96.0 % (calcd.on anh.substance)
- Water:3.0 to 7.0 %
Frequently Bought Together