Order Entry
Netherlands
ContactUsLinkComponent
 
(Diethoxyphosphoryl)methyl-4-methylbenzenesulphonate ≥97.0% (by GC)
  TCIAD4968-25G
 :  TCI
 :  
(Diethoxyphosphoryl)methyl-4-methylbenzenesulphonate ≥97.0% (by GC)
  TCIAD4968-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :137°C
 :322.3100 g/mol
 :Refrigerator
 :31618-90-3
 :C₁₂H₁₉O₆PS
 :MFCD03412078