2,6-Dibromo-N,N'-di-n-octyl-1,8:4,5-naphthalenetetracarboxdiimide ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:926643-78-9
:
Formula:C₃₀H₃₆Br₂N₂O₄
MDL Number:MFCD29472527
:648.44 g/mol
Specification Test Results
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together