Order Entry
Netherlands
ContactUsLinkComponent
2,4-Dimethoxyphenyl isothiocyanate ≥98.0% (by GC)
Corrosive
Irritant
2,4-Dimethoxyphenyl isothiocyanate ≥98.0% (by GC)
Supplier:  TCI
Certificates
Corrosive
Irritant
2,4-Dimethoxyphenyl isothiocyanate ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    52.0 to 56.0 deg-C

Specification Test Results

Frequently Bought Together