2,7-Di(1-pyrenyl)-9,9'-spirobi[9H-fluorene] ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:886456-80-0
:
Formula:C₅₇H₃₂
MDL Number:MFCD12022456
:716.88 g/mol
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Frequently Bought Together