Order Entry
Netherlands
ContactUsLinkComponent
 
Dimethyl 4-Hydroxy-2,6-pyridinedicarboxylate monohydrate ≥98.0% (by GC)
 :  TCI
 
Dimethyl 4-Hydroxy-2,6-pyridinedicarboxylate monohydrate ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Water:
    6.0 to 10.0 %

 

Frequently Bought Together