
2,4-Xylyl isothiocyanate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:39842-01-8
:
Formula:C₉H₉NS
MDL Number:MFCD00022054
:163.24 g/mol
Boiling Pt:263°C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White or Colorless to Light orange to Yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together