Order Entry
Netherlands
ContactUsLinkComponent
6,7-Dimethyl-1-tetralone ≥98.0% (by GC)
6,7-Dimethyl-1-tetralone ≥98.0% (by GC)
Catalog # TCIAD4908-1G
Supplier:  TCI
CAS Number:  
undefined
6,7-Dimethyl-1-tetralone ≥98.0% (by GC)
Catalog # TCIAD4908-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    46.0 to 50.0 deg-C

Specification Test Results

About this item

 :
 :174.24 g/mol
Boiling Pt:130°C
CAS Number:19550-57-3
Formula:C₁₂H₁₄O
MDL Number:MFCD00156729